Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411751

Sigma-Aldrich

Anti-MBNL3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CHCR, FLJ11316, MBLX, MBLX39, MBXL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBNL3(55796)

Immunogen

MBNL3 (AAH42090.1, 1 a.a. ~ 258 a.a) full-length human protein.

Sequence
MFAQQMQLMLQNAQMSSLGSFPMTPSIPANPPMAFNPYIPHPGMGLVPAELVPNTPVLIPGNPPLAMPGAVGPKLMRSDKLEVCREFQRGNCTRGENDCRYAHPTDASMIEASDNTVTICMDYIKGRCSREKCKYFHPPAHLQARLKAAHHQMNHSAASAMALQPGTLQLIPKRSALEKPNGATPVFNPTVFHCQQALTNLQLPQPAFIPAGPILCMAPASNIVPMMHGATPTTVSAATTPATSVPFAAPTTGNQLKF

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadiya M Teplyuk et al.
EMBO molecular medicine, 8(3), 268-287 (2016-02-18)
MicroRNA-10b (miR-10b) is a unique oncogenic miRNA that is highly expressed in all GBM subtypes, while absent in normal neuroglial cells of the brain. miR-10b inhibition strongly impairs proliferation and survival of cultured glioma cells, including glioma-initiating stem-like cells (GSC).
Melissa N Hinman et al.
Disease models & mechanisms, 14(6) (2021-06-15)
The muscleblind RNA-binding proteins (MBNL1, MBNL2 and MBNL3) are highly conserved across vertebrates and are important regulators of RNA alternative splicing. Loss of MBNL protein function through sequestration by CUG or CCUG RNA repeats is largely responsible for the phenotypes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service