Skip to Content
Merck
All Photos(3)

Key Documents

SAB1411438

Sigma-Aldrich

Anti-RPAP3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ21908, spag

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Check Cart for Availability


Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$660.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 71.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPAP3(79657)

General description

RNA polymerase II associated protein 3 (RPAP3) gene encodes an RNA polymerase II-associated protein. The encoded protein may function in transcriptional regulation and may also regulate apoptosis. Alternatively spliced transcript variants have been described. (provided by RefSeq). The protein consists of tetratricopeptide repeat (TPR) domain.

Immunogen

RPAP3 (AAH56415.1, 1 a.a. ~ 631 a.a) full-length human protein.

Sequence
MTSANKAIELQLQVKQNAEELQDFMRDLENWEKDIKQKDMELRRQNGVPEENLPPIRNGNFRKKKKGKAKESSKKTREENTKNRIKSYDYEAWAKLDVDRILDELDKDDSTHESLSQESESEEDGIHVDSQKALVLKEKGNKYFKQGKYDEAIDCYTKGMDADPYNPVLPTNRASAYFRLKKFAVAESDCNLAVALNRSYTKAYSRRGAARFALQKLEEAKKDYERVLELEPNNFEATNELRKISQALASKENSYPKEADIVIKSTEGERKQIEAQQNKQQAISEKDRGNGFFKEGKYERAIECYTRGIAADGANALLPANRAMAYLKIQKYEEAEKDCTQAILLDGSYSKAFARRGTARTFLGKLNEAKQDFETVLLLEPGNKQAVTELSKIKKKPLKKVIIEETGNLIQTIDVPDSTTAAAPENNPINLANVIAATGTTSKKNSSQDDLFPTSDTPRAKVLKIEEVSDTSSLQPQASLKQDVCQSYSEKMPIEIEQKPAQFATTVLPPIPANSFQLESDFRQLKSSPDMLYQYLKQIEPSLYPKLFQKNLDPDVFNQIVKILHDFYIEKEKPLLIFEILQRLSELKRFDMAVMFMSETEKKIARALFNHIDKSGLKDSSVEELKKRYGG

Application

Anti-RPAP3 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

RNA polymerase II associated protein 3 (RPAP3) acts as a cell death enhancer. The protein may modulate nuclear factor-kappa B (NF-κb) pathway in apoptosis, which is induced by anti-cancer chemotherapeutic agents. It functions as a Monad binding protein. This protein regulates apoptosis and small nucleolar ribonucleoparticule assembly process.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1 H, 15 N and 13 C resonance assignments of the two TPR domains from the human RPAP3 protein
Chagot M, et al.
Biomolecular NMR Assignments, 9(1), 99-102 (2015)
RPAP3 interacts with Reptin to regulate UV-induced phosphorylation of H2AX and DNA damage
Ni L, et al.
Journal of Cellular Biochemistry, 106(5), 920-928 (2009)
RPAP3 splicing variant isoform 1 interacts with PIH1D1 to compose R2TP complex for cell survival
Yoshida M, et al.
Biochemical and Biophysical Research Communications, 430(1), 320-324 (2013)
PIH1D1 interacts with mTOR complex 1 and enhances ribosome RNA transcription
Kamano Y, et al.
Febs Letters, 587(20), 3303-3308 (2013)
RPAP3 enhances cytotoxicity of doxorubicin by impairing NF-kappa B pathway
Shimada K, et al.
Biochemical and Biophysical Research Communications, 404(4), 910-914 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service