Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411292

Sigma-Aldrich

Anti-BMPR2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Estimated to ship on18 April 2025



Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$660.00


Estimated to ship on18 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BMPR2(659)

General description

This gene encodes a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs, which are members of the TGF-beta superfamily. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in this gene have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease. (provided by RefSeq)

Immunogen

BMPR2 (NP_001195.2, 1 a.a. ~ 1038 a.a) full-length human protein.

Sequence
MTSSLQRPWRVPWLPWTILLVSTAAASQNQERLCAFKDPYQQDLGIGESRISHENGTILCSKGSTCYGLWEKSKGDINLVKQGCWSHIGDPQECHYEECVVTTTPPSIQNGTYRFCCCSTDLCNVNFTENFPPPDTTPLSPPHSFNRDETIIIALASVSVLAVLIVALCFGYRMLTGDRKQGLHSMNMMEAAASEPSLDLDNLKLLELIGRGRYGAVYKGSLDERPVAVKVFSFANRQNFINEKNIYRVPLMEHDNIARFIVGDERVTADGRMEYLLVMEYYPNGSLCKYLSLHTSDWVSSCRLAHSVTRGLAYLHTELPRGDHYKPAISHRDLNSRNVLVKNDGTCVISDFGLSMRLTGNRLVRPGEEDNAAISEVGTIRYMAPEVLEGAVNLRDCESALKQVDMYALGLIYWEIFMRCTDLFPGESVPEYQMAFQTEVGNHPTFEDMQVLVSREKQRPKFPEAWKENSLAVRSLKETIEDCWDQDAEARLTAQCAEERMAELMMIWERNKSVSPTVNPMSTAMQNERNLSHNRRVPKIGPYPDYSSSSYIEDSIHHTDSIVKNISSEHSMSSTPLTIGEKNRNSINYERQQAQARIPSPETSVTSLSTNTTTTNTTGLTPSTGMTTISEMPYPDETNLHTTNVAQSIGPTPVCLQLTEEDLETNKLDPKEVDKNLKESSDENLMEHSLKQFSGPDPLSSTSSSLLYPLIKLAVEATGQQDFTQTANGQACLIPDVLPTQIYPLPKQQNLPKRPTSLPLNTKNSTKEPRLKFGSKHKSNLKQVETGVAKMNTINAAEPHVVTVTMNGVAGRNHSVNSHAATTQYANGTVLSGQTTNIVTHRAQEMLQNQFIGEDTRLNINSSPDEHEPLLRREQQAGHDEGVLDRLVDRRERPLEGGRTNSNNNNSNPCSEQDVLAQGVPSTAADPGPSKPRRAQRPNSLDLSATNVLDGSSIQIGESTQDGKSGSGEKIKKRVKTPYSLKRWRPSTWVISTESLDCEVNNNGSNRAVHSKSSTAVYLAEGGTATTMVSKDIGMNCL

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service