Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411232

Sigma-Aldrich

Anti-CCL4 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ACT2, AT744.1, G-26, LAG1, MGC104418, MGC126025, MGC126026, MIP-1-beta, MIP1B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Check Cart for Availability


Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$660.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 10.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL4(6351)

Related Categories

General description

Both MIP-1α and MIP-1β (macrophage inflammatory proteins-1α and β) are structurally and functionally related CC chemokines. They are secreted by activated human monocytes and peripheral blood lymphocytes. The gene encoding this protein is localized on human chromosome 17q12.

Immunogen

CCL4 (NP_002975.1, 1 a.a. ~ 92 a.a) full-length human protein.

Sequence
MKLCVTVLSLLMLVAAFCSLALSAPMGSDPPTACCFSYTARKLPRNFVVDYYETSSLCSQPAVVFQTKRSKQVCADPSESWVQEYVYDLELN

Biochem/physiol Actions

MIP-1α and MIP-1β (macrophage inflammatory proteins-1α and β) participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells e.g. macrophages, lymphocytes and natural killer (NK) cells. While both MIP-1α and MIP-1β exert similar effects on monocytes their effect on lymphocytes differ; with MIP-1α selectively attracting CD8+ lymphocytes and MIP-1β (also known as C-C motif chemokine ligand 4) selectively attracting CD4+ lymphocytes. Additionally, MIP-1α and MIP-1β have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of human macrophage inflammatory proteins 1alpha and 1beta as a native secreted heterodimer.
Guan E
The Journal of Biological Chemistry, 276(15), 12404-12409 (2001)
CCL3L1 and CCL4L1 chemokine genes are located in a segmental duplication at chromosome 17q12.
Modi WS
Genomics, 83(4), 735-738 (2004)
MicroRNA-125b modulates inflammatory chemokine CCL4 expression in immune cells and its reduction causes CCL4 increase with age.
Cheng NL
Aging Cell, 14(2), 200-208 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service