Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411220

Sigma-Aldrich

Anti-S100A9 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 13.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A9(6280)

Related Categories

General description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis. (provided by RefSeq)

Immunogen

S100A9 (NP_002956.1, 1 a.a. ~ 114 a.a) full-length human protein.

Sequence
MTCKMSQLERNIETIINTFHQYSVKLGHPDTLNQGEFKELVRKDLQNFLKKENKNEKVIEHIMEDLDTNADKQLSFEEFIMLMARLTWASHEKMHEGDEGPGHHHKPGLGEGTP

Biochem/physiol Actions

S100A9 (S100 calcium binding protein A9) has pro-inflammatory action in acute and chronic inflammation. In addition to inflammatory action, it exhibits cytokine-like activities for the activation of the receptor for advanced glycation end products (RAGE) and Toll-like receptor 4-dependent signaling cascades. S100A9 stimulates neutrophil chemotaxis. It stimulates shedding of L-selectin, up-regulates and activates Mac-1 which finally induce neutrophil adhesion to fibrinogen in vitro.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carle Ryckman et al.
Journal of immunology (Baltimore, Md. : 1950), 170(6), 3233-3242 (2003-03-11)
S100A8 and S100A9 are small calcium-binding proteins that are highly expressed in neutrophil and monocyte cytosol and are found at high levels in the extracellular milieu during inflammatory conditions. Although reports have proposed a proinflammatory role for these proteins, their
Karen Oliva et al.
The Journal of endocrinology, 218(2), 165-178 (2013-05-28)
Gestational diabetes mellitus (GDM) is a significant risk factor for the type 2 diabetes epidemic in many populations. Maternal adipose tissue plays a central role in the pathophysiology of GDM. Thus, the aim of this study was to determine the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service