Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410861

Sigma-Aldrich

Anti-OPRL1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KOR-3, MGC34578, NOCIR, OOR, ORL1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OPRL1(4987)

General description

Opioid related nociceptin receptor 1 (OPRL1) is mapped to human chromosome 20q13.33. The gene codes for the nociceptin/orphanin FQ receptor (NOP-R), which is a G protein-coupled receptor expressed at various levels all over the brain and spinal cord. OPRL1 belongs to the opioid receptor family.
The protein encoded by this gene is a G protein-coupled receptor whose expression can be induced by phytohemagglutinin. The encoded integral membrane protein is a receptor for the 17 aa neuropeptide nociceptin/orphanin FQ. This gene may be involved in the regulation of numerous brain activities, particularly instinctive and emotional behaviors. A promoter for this gene also functions as a promoter for another gene, regulator of G-protein signalling 19 (RGS19), located on the opposite strand. Two transcript variants encoding the same protein have been found for this gene. (provided by RefSeq)

Immunogen

OPRL1 (NP_000904.1, 1 a.a. ~ 370 a.a) full-length human protein.

Sequence
MEPLFPAPFWEVIYGSHLQGNLSLLSPNHSLLPPHLLLNASHGAFLPLGLKVTIVGLYLAVCVGGLLGNCLVMYVILRHTKMKTATNIYIFNLALADTLVLLTLPFQGTDILLGFWPFGNALCKTVIAIDYYNMFTSTFTLTAMSVDRYVAICHPIRALDVRTSSKAQAVNVAIWALASVVGVPVAIMGSAQVEDEEIECLVEIPTPQDYWGPVFAICIFLFSFIVPVLVISVCYSLMIRRLRGVRLLSGSREKDRNLRRITRLVLVVVAVFVGCWTPVQVFVLAQGLGVQPSSETAVAILRFCTALGYVNSCLNPILYAFLDENFKACFRKFCCASALRRDVQVSDRVRSIAKDVALACKTSETVPRPA

Biochem/physiol Actions

Opioid related nociceptin receptor 1 (OPRL1) plays a vital role in alcohol and other drug reward pathways. The encoded protein is involved in regulating tolerance and behavioral responses to morphine. Phosphorylation of NOPR at serine 363 by G protein-coupled receptor kinase 3 (GRK3) facilitates arrestin-dependent Jun amino-terminal kinases (JNK) mitogen-activated protein kinase (MAPK) signaling. Mutation in the gene is associated with the risk to develop opiate addiction.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tineke E Buffart et al.
Virchows Archiv : an international journal of pathology, 455(3), 213-223 (2009-08-22)
DNA copy-number gains of chromosomes 8q, 13q, and 20q are frequently observed in gastric cancers. Moreover gain of chromosome 20q has been associated with lymph node metastasis. The aim of this study was to correlate DNA copy-number changes of individual
Judith A Briant et al.
Psychiatric genetics, 20(2), 65-72 (2009-12-25)
The OPRL1 gene encodes the nociceptin/orphanin FQ receptor, which plays a role in regulating tolerance and behavioral responses to morphine. However, there is limited information on whether variants of OPRL1 are associated with vulnerability to develop opiate addiction. In this
Xiaoling Xuei et al.
Addiction biology, 13(1), 80-87 (2007-10-04)
Recent studies in animal models have shown that the nociceptin system, comprising nociceptin (or OFQ/N, encoded by PNOC) and the nociceptin receptor (an opioid receptor-like protein encoded by OPRL1), may be involved in alcohol and other drug reward pathways. To
Nancy R Zhang et al.
The Journal of biological chemistry, 287(50), 42019-42030 (2012-10-23)
We determined the role of carboxyl-terminal regulation of NOPR (nociceptin, orphanin FQ receptor) signaling and function. We mutated C-terminal serine and threonine residues and examined their role in NOPR trafficking, homologous desensitization, and arrestin-dependent MAPK signaling. The NOPR agonist, nociceptin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service