Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410792

Sigma-Aldrich

Anti-MOG antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC26137, MOGIG-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Check Cart for Availability


Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$660.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 32.56 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MOG(4340)

General description

The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintenance of the myelin sheath and in cell-cell communication. Alternatively spliced transcript variants encoding different isoforms have been identified. (provided by RefSeq)

Immunogen

MOG (AAH35938.1, 1 a.a. ~ 295 a.a) full-length human protein.

Sequence
MASLSRPSLPSCLCSFLLLLLLQVSSSYAGQFRVIGPRHPIRALVGDEVELPCRISPGKNATGMEVGWYRPPFSRVVHLYRNGKDQDGDQAPEYRGRTELLKDAIGEGKVTLRIRNVRFSDEGGFTCFFRDHSYQEEAAMELKVEDPFYWVSPGVLVLLAVLPVLLLQITVGLVFLCLQYRLRGKLRAEIENLHRTFDPHFLRVPCWKITLFVIVPVLGPLVALIICYNWLHRRLAGQFLEELRKFSSLCYKQRIKSQERETEATRGRGGLLRDHIPRGKEELESLGGGKTPPGR

Biochem/physiol Actions

Myelin/oligodendrocyte glycoprotein (MOG) is a major component of the central nervous system. It is involved in several cellular processes including cellular adhesion, in the regulation of oligodendrocyte microtubule stability and facilitates the interactions between myelin and the immune system. It has been reported that MOG may be involved in the completion and/or maintenance of the myelin sheath, and in cell-cell communication. It has also been observed that mutation or polymorphisms in MOG gene could cause multiple sclerosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B della Gaspera et al.
European journal of biochemistry, 258(2), 478-484 (1999-01-05)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the mammalian central nervous system, is located on the surface of the oligodendrocyte plasma membrane and the outermost lamellae of mature myelin; it is expressed during the latter steps of myelinogenesis. It has
D Pham-Dinh et al.
Genomics, 29(2), 345-352 (1995-09-20)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the central nervous system localized on the outermost lamellae of mature myelin, is a member of the immunoglobulin superfamily. We report here the organization of the human MOG gene, which spans approximately 17
T G Johns et al.
Journal of neurochemistry, 72(1), 1-9 (1999-01-14)
Myelin oligodendrocyte glycoprotein (MOG) is a quantitatively minor component of CNS myelin whose function remains relatively unknown. As MOG is an autoantigen capable of producing a demyelinating multiple sclerosis-like disease in mice and rats, much of the research directed toward
D Pham-Dinh et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(17), 7990-7994 (1993-09-01)
Myelin/oligodendrocyte glycoprotein (MOG) is found on the surface of myelinating oligodendrocytes and external lamellae of myelin sheaths in the central nervous system, and it is a target antigen in experimental autoimmune encephalomyelitis and multiple sclerosis. We have isolated bovine, mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service