Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410595

Sigma-Aldrich

Anti-BRF1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BRF, FLJ42674, FLJ43034, GTF3B, MGC105048, TAF3B2, TAF3C, TAFIII90, TF3B90

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 51.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRF1(2972)

General description

This gene encodes one of the three subunits of the RNA polymerase III transcription factor complex. This complex plays a central role in transcription initiation by RNA polymerase III on genes encoding tRNA, 5S rRNA, and other small structural RNAs. The gene product belongs to the TF2B family. Two alternatively spliced variants encoding different isoforms, that function at different promoters transcribed by RNA polymerase III, have been identified. Other transcript variants are possible, but their full-length natures have not been completely characterized. (provided by RefSeq)

Immunogen

BRF1 (NP_663718.1, 1 a.a. ~ 473 a.a) full-length human protein.

Sequence
MTALRLLQRMKRDWMHTGRRPSGLCGAALLVAARMHDFRRTVKEVISVVKVCESTLRKRLTEFEDTPTSQLTIDEFMKIDLEEECDPPSYTAGQRKLRMKQLEQVLSKKLEEVEGEISSYQDAIEIELENSRPKAKGGLASLAKDGSTEDTASSLCGEEDTEDEELEAAASHLNKDLYRELLGGAPGSSEAAGSPEWGGRPPALGSLLDPLPTAASLGISDSIRECISSQSSDPKDASGDGELDLSGIDDLEIDRYILNESEARVKAELWMRENAEYLREQREKEARIAKEKELGIYKEHKPKKSCKRREPIQASTAREAIEKMLEQKKISSKINYSVLRGLSSAGGGSPHREDAQPEHSASARKLSRRRTPASRSGADPVTSVGKRLRPLVSTQPAKKVATGEALLPSSPTLGAEPARPQAVLVESGPVSYHADEEADEEEPDEEDGEPCVSALQMMGSNDYGCDGDEDDGY

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service