Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410583

Sigma-Aldrich

Anti-SLC25A6 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AAC3, ANT3, ANT3Y, MGC17525

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Estimated to ship on20 April 2025



Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Estimated to ship on20 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 32.9 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC25A6(293)

General description

This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene. (provided by RefSeq)

Immunogen

SLC25A6 (AAH14775.1, 1 a.a. ~ 298 a.a) full-length human protein.

Sequence
MTEQAISFAKDFLAGGIAAAISKTAVAPIERVKLLLQVQHASKQIAADKQYKGIVDCIVRIPKEQGVLSFWRGNLANVIRYFPTQALNFAFKDKYKQIFLGGVDKHTQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKSGTEREFRGLGDCLVKITKSDGIRGLYQGFSVSVQGIIIYRAAYFGVYDTAKGMLPDPKNTHIVVSWMIAQTVTAVAGVVSYPFDTVRRRMMMQFGRKGADIMYTGTVDCWRKIFRDEGGKAFFKGAWSNVLRGMGGAFVLVLYDELKKVI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service