Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410410

Sigma-Aldrich

Anti-SULF1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ30905, FLJ38022, FLJ41750, HSULF-1, KIAA1077, SULF-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$777.00

$777.00


Check Cart for Availability


Select a Size

Change View
100 μG
$777.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$777.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 101 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULF1(23213)

General description

Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 [PubMed 16192265]).[supplied by OMIM

Immunogen

SULF1 (NP_055985.1, 1 a.a. ~ 871 a.a) full-length human protein.

Sequence
MKYSCCALVLAVLGTELLGSLCSTVRSPRFRGRIQQERKNIRPNIILVLTDDQDVELGSLQVMNKTRKIMEHGGATFINAFVTTPMCCPSRSSMLTGKYVHNHNVYTNNENCSSPSWQAMHEPRTFAVYLNNTGYRTAFFGKYLNEYNGSYIPPGWREWLGLIKNSRFYNYTVCRNGIKEKHGFDYAKDYFTDLITNESINYFKMSKRMYPHRPVMMVISHAAPHGPEDSAPQFSKLYPNASQHITPSYNYAPNMDKHWIMQYTGPMLPIHMEFTNILQRKRLQTLMSVDDSVERLYNMLVETGELENTYIIYTADHGYHIGQFGLVKGKSMPYDFDIRVPFFIRGPSVEPGSIVPQIVLNIDLAPTILDIAGLDTPPDVDGKSVLKLLDPEKPGNRFRTNKKAKIWRDTFLVERGKFLRKKEESSKNIQQSNHLPKYERVKELCQQARYQTACEQPGQKWQCIEDTSGKLRIHKCKGPSDLLTVRQSTRNLYARGFHDKDKECSCRESGYRASRSQRKSQRQFLRNQGTPKYKPRFVHTRQTRSLSVEFEGEIYDINLEEEEELQVLQPRNIAKRHDEGHKGPRDLQASSGGNRGRMLADSSNAVGPPTTVRVTHKCFILPNDSIHCERELYQSARAWKDHKAYIDKEIEALQDKIKNLREVRGHLKRRKPEECSCSKQSYYNKEKGVKKQEKLKSHLHPFKEAAQEVDSKLQLFKENNRRRKKERKEKRRQRKGEECSLPGLTCFTHDNNHWQTAPFWNLGSFCACTSSNNNTYWCLRTVNETHNFLFCEFATGFLEYFDMNTDPYQLTNTVHTVERGILNQLHVQLMELRSCQGYKQCNPRPKNLDVGNKDGGSYDLHRGQLWDGWEG

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan J Weiss et al.
Nature chemical biology, 17(6), 684-692 (2021-04-14)
Heparan sulfate (HS) proteoglycans bind extracellular proteins that participate in cell signaling, attachment and endocytosis. These interactions depend on the arrangement of sulfated sugars in the HS chains generated by well-characterized biosynthetic enzymes; however, the regulation of these enzymes is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service