Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410294

Sigma-Aldrich

Anti-DFFB antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CAD, CPAN, DFF-40, DFF2, DFF40

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 39.1 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DFFB(1677)

General description

Apoptosis is a cell death process that removes toxic and/or useless cells during mammalian development. The apoptotic process is accompanied by shrinkage and fragmentation of the cells and nuclei and degradation of the chromosomal DNA into nucleosomal units. DNA fragmentation factor (DFF) is a heterodimeric protein of 40-kD (DFFB) and 45-kD (DFFA) subunits. DFFA is the substrate for caspase-3 and triggers DNA fragmentation during apoptosis. DFF becomes activated when DFFA is cleaved by caspase-3. The cleaved fragments of DFFA dissociate from DFFB, the active component of DFF. DFFB has been found to trigger both DNA fragmentation and chromatin condensation during apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene but the biological validity of these variants has not been determined. (provided by RefSeq)

Immunogen

DFFB (NP_004393.1, 1 a.a. ~ 338 a.a) full-length human protein.

Sequence
MLQKPKSVKLRALRSPRKFGVAGRSCQEVLRKGCLRFQLPERGSRLCLYEDGTELTEDYFPSVPDNAELVLLTLGQAWQGYVSDIRRFLSAFHEPQVGLIQAAQQLLCDEQAPQRQRLLADLLHNVSQNIAAETRAEDPPWFEGLESRFQSKSGYLRYSCESRIRSYLREVSSYPSTVGAEAQEEFLRVLGSMCQRLRSMQYNGSYFDRGAKGGSRLCTPEGWFSCQGPFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service