Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410181

Sigma-Aldrich

Anti-RBPJL antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

RBP-L, RBPSUHL, SUH, SUHL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBPJL(11317)

General description

In mouse, recombining binding protein L (RBP-L) is a transcription factor that binds to DNA sequences almost identical to that bound by the Notch receptor signalling pathway transcription factor RBP-J. However, unlike RBP-J, RBP-L does not interact with Notch receptors. RBP-L has been shown to activate transcription in concert with Epstein-Barr virus nuclear antigen-2 (EBNA2). The protein encoded by this gene is similar in sequence to the mouse RPB-L protein and Drosophila suppressor of hairless protein. (provided by RefSeq)

Immunogen

RBPJL (NP_055091.2, 1 a.a. ~ 517 a.a) full-length human protein.

Sequence
MDPAGAADPSVPPNPLTHLSLQDRSEMQLQSEADRRSLPGTWTRSSPEHTTILRGGVRRCLQQQCEQTVRILHAKVAQKSYGNEKRFFCPPPCVYLSGPGWRVKPGQDQAHQAGETGPTVCGYMGLDSASGSATETQKLNFEQQPDSREFGCAKTLYISDADKRKHFRLVLRLVLRGGRELGTFHSRLIKVISKPSQKKQSLKNTDLCISSGSKVSLFNRLRSQTVSTRYLSVEDGAFVASARQWAAFTLHLADGHSAQGDFPPREGYVRYGSLVQLVCTVTGITLPPMIIRKVAKQCALLDVDEPISQLHKCAFQFPGSPPGGGGTYLCLATEKVVQFQASPCPKEANRALLNDSSCWTIIGTESVEFSFSTSLACTLEPVTPVPLISTLELSGGGDVATLELHGENFHAGLKVWFGDVEAETMYRSPRSLVCVVPDVAAFCSDWRWLRAPITIPMSLVRADGLFYPSAFSFTYTPEYSVRPGHPGVPEPATDADALLESIHQEFTRTNFHLFIQT

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service