Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409898

Sigma-Aldrich

Monoclonal Anti-TTF2 antibody produced in mouse

clone 1E4, ascites fluid, solution

Synonym(s):

HuF2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
200 μL
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1E4, monoclonal

form

solution

mol wt

antigen 39.38 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTF2(8458)

General description

This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing. (provided by RefSeq)

Immunogen

TTF2 (NP_003585, 385 a.a. ~ 509 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QFPDRSVQRKVSPASGVSKKVEPSDPVARRVYLTTQLKQKKSTLASVNIQALPDKGQKLIKQIQELEEVLSGLTLSPEQGTNEKSNSQVPQQSHFTKTTTGPPHLVPPQPLPRRGTQPVGSLELK

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service