Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409854

Sigma-Aldrich

Monoclonal Anti-CAMK2D antibody produced in mouse

clone 1B11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CAMKD, DKFZp686G23119, DKFZp686I2288, MGC44911

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B11, monoclonal

form

buffered aqueous solution

mol wt

antigen 37.73 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK2D(817)

Immunogen

CAMK2D (AAH32784, 301 a.a. ~ 410 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KGAILTTMLATRNFSAAKSLLKKPDGVKESTESSNTTIEDEDVKARKQEIIKVTEQLIEAINNGDFEAYTKICDPGLTAFEPEALGNLVEGMDFHRFYFENALSKSNKPI

Biochem/physiol Actions

As a subunit of the calcium/calmodulin dependent protein kinase II complex, calcium/calmodulin dependent protein kinase II complex δ (CAMK2D) modulates the functions of ion channels and Ca2+ handling proteins. It also regulates the phosphorylation and acetylation of histones. It has been shown to be overexpressed in the myocardium of diabetic patients.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorna Daniels et al.
Heart failure reviews, 20(5), 589-600 (2015-07-23)
Diabetes mellitus (DM) is an increasing epidemic that places a significant burden on health services worldwide. The incidence of heart failure (HF) is significantly higher in diabetic patients compared to non-diabetic patients. One underlying mechanism proposed for the link between
Salma Awad et al.
The Journal of pathology, 235(4), 606-618 (2014-11-26)
Heart failure is associated with the reactivation of a fetal cardiac gene programme that has become a hallmark of cardiac hypertrophy and maladaptive ventricular remodelling, yet the mechanisms that regulate this transcriptional reprogramming are not fully understood. Using mice with
Samuel Sossalla et al.
Circulation research, 107(9), 1150-1161 (2010-09-04)
Heart failure (HF) is known to be associated with increased Ca(2+)/calmodulin-dependent protein kinase (CaMK)II expression and activity. There is still controversial discussion about the functional role of CaMKII in HF. Moreover, CaMKII inhibition has never been investigated in human myocardium.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service