Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409827

Sigma-Aldrich

Anti-KIAA0319L antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA1837, PKD1-like, PP791, RP4-765A10.3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$660.00

$660.00


Estimated to ship on01 May 2025



Select a Size

Change View
50 μG
$660.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$660.00


Estimated to ship on01 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

KIAA0319-like (KIAA0319L) protein is localized in the nucleus and cytoplasm. This protein is a type-I transmembrane protein with five immunoglobulin-like domains, known as polycystic kidney disease (PKD) domains present in the ectodomain. The KIAA0319L gene is mapped on the human chromosome at 1p34.3.

Immunogen

KIAA0319L (AAH14530.1, 1 a.a. ~ 1049 a.a) full-length human protein.

Sequence
MEKRLGVKPNPASWILSGYYWQTSAKWLRSLYLFYTCFCFSVLWLSTDASESRCQQGKTQFGVGLRSGGENHLWLLEGTPSLQSCLAACCQDSACHVFWWLEGMCIQADCSRPQSCRAFRTHSSNSMLVFLKKFQTADDLGFLPEDDVPHLLGLGWNWASWRQSPPRAALRPAVSSSDQQSLIRKLQKRGSPSDVVTPIVTQHSKVNDSNELGGLTTSGSAEVHKAITISSPLTTDLTAELSGGPKNVSVQPEISEGLATTPSTQQVKSSEKTQIAVPQPVAPSYSYATPTPQASFQSTSAPYPVIKELVVSAGESVQITLPKNEVQLNAYVLQEPPKGETYTYDWQLITHPRDYSGEMEGKHSQILKLSKLTPGLYEFKVIVEGQNAHGEGYVNVTVKPEPRKNRPPIAIVSPQFQEISLPTTSTVIDGSQSTDDDKIVQYHWEELKGPLREEKISEDTAILKLSKLVPGNYTFSLTVVDSDGATNSTTANLTVNKAVDYPPVANAGPNQVITLPQNSITLFGNQSTDDHGITSYEWSLSPSSKGKVVEMQGVRTPTLQLSAMQEGDYTYQLTVTDTIGQQATAQVTVIVQPENNKPPQADAGPDKELTLPVDSTTLDGSKSSDDQKIISYLWEKTQGPDGVQLENANSSVATVTGLQVGTYVFTLTVKDERNLQSQSSVNVIVKEEINKPPIAKITGNVVITLPTSTAELDGSKSSDDKGIVSYLWTRDEGSPAAGEVLNHSDHHPILFLSNLVEGTYTFHLKVTDAKGESDTDRTTVEVKPDPRKNNLVEIILDINVSQLTERLKGMFIRQIGVLLGVLDSDIIVQKIQPYTEQSTKMVFFVQNEPPHQIFKGHEVAAMLKSELRKQKADFLIFRALEVNTVTCQLNCSDHGHCDSFTKRCICDPFWMENFIKVQLRDGDSNCEWSVLYVIIATFVIVVALGILSWTVICCCKRQKGKPKRKSKYKILDATDQESLELKPTSRAGIKQKGLLLSSSLMHSESELDSDDAIFTWPDREKGKLLHGQNGSVPNGQTPLKARSPREEIL

Biochem/physiol Actions

KIAA0319-like (KIAA0319L) protein is involved in neuronal migration in the developing brain. It plays a role in learning and cognition. KIAA0319L works as a receptor for adeno-associated virus infection. KIAA0319L gene is considered a dyslexia susceptibility gene that is associated with developmental dyslexia, a common childhood learning disorder. Overexpression of the KIAA0319L gene is observed in peripheral blood cells from patients with systemic lupus erythematosus (SLE). KIAA0319L protein is linked with polycystic kidney disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guihong Song et al.
Reproductive biomedicine online, 30(3), 275-280 (2015-01-19)
KIAA0319L, PXK and JAZF1 gene polymorphisms were investigated to determine whether they conferred susceptibility to unexplained recurrent pregnancy loss (URPL) in a group of Chinese Han patients. Genotyping and sequencing of the single nucleotide polymorphisms (SNP) rs2275247(A/G) in KIAA0319L, rs2176082(C/T)
Luiz G Guidi et al.
Cerebral cortex (New York, N.Y. : 1991), 27(12), 5831-5845 (2017-10-19)
Developmental dyslexia is a neurodevelopmental disorder that affects reading ability caused by genetic and non-genetic factors. Amongst the susceptibility genes identified to date, KIAA0319 is a prime candidate. RNA-interference experiments in rats suggested its involvement in cortical migration but we
An essential receptor for adeno-associated virus infection.
Pillay S
Nature, 530, 108-108 (2016)
Antonio Velayos-Baeza et al.
Human molecular genetics, 17(6), 859-871 (2007-12-08)
The KIAA0319 gene has been recently associated with developmental dyslexia and shown to be involved in neuronal migration. The deduced KIAA0319 protein contains several polycystic kidney disease (PKD) domains which may mediate the interaction between neurons and glial fibres during
Jillian M Couto et al.
Journal of neurogenetics, 22(4), 295-313 (2008-12-17)
A locus on chromosome 1p34-36 (DYX8) has been linked to developmental dyslexia or reading disabilities (RD) in three independent samples. In the current study, we investigated a candidate gene KIAA0319-Like (KIAA0319L) within DYX8, as it is homologous to KIAA0319, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service