Skip to Content
Merck
All Photos(1)

Key Documents

SAB1409548

Sigma-Aldrich

Anti-MAN1C1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HMIC, MAN1A3, MAN1C, pp6318

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$660.00

$660.00


Check Cart for Availability


Select a Size

Change View
50 μG
$660.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$660.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 70.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAN1C1(57134)

Immunogen

MAN1C1 (NP_065112.1, 1 a.a. ~ 630 a.a) full-length human protein.

Sequence
MLMRKVPGFVPASPWGLRLPQKFLFLLFLSGLVTLCFGALFLLPHSSRLKRLFLAPRTQQPGLEVVAEIAGHAPAREQEPPPNPAPAAPAPGEDDPSSWASPRRRKGGLRRTRPTGPREEATAARGNSIPASRPGDEGVPFRFDFNAFRSRLRHPVLGTRADESQEPQSQVRAQREKIKEMMQFAWQSYKRYAMGKNELRPLTKDGYEGNMFGGLSGATVIDSLDTLYLMELKEEFQEAKAWVGESFHLNVSGEASLFEVNIRYIGGLLSAFYLTGEEVFRIKAIRLGEKLLPAFNTPTGIPKGVVSFKSGNWGWATAGSSSILAEFGSLHLEFLHLTELSGNQVFAEKVRNIRKVLRKIEKPFGLYPNFLSPVSGNWVQHHVSVGGLGDSFYEYLIKSWLMSGKTDMEAKNMYYEALEAIETYLLNVSPGGLTYIAEWRGGILDHKMGHLACFSGGMIALGAEDAKEEKRAHYRELAAQITKTCHESYARSDTKLGPEAFWFNSGREAVATQLSESYYILRPEVVESYMYLWRQTHNPIYREWGWEVVLALEKYCRTEAGFSGIQDVYSSTPNHDNKQQSFFLAETLKYLYLLFSEDDLLSLEDWVFNTEAHPLPVNHSDSSGRAWGRH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service