Skip to Content
Merck
All Photos(1)

Key Documents

SAB1409385

Sigma-Aldrich

Anti-NUP98 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADIR2, NUP196, NUP96

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
50 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 69.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUP98(4928)

General description

Signal-mediated nuclear import and export proceed through the nuclear pore complex (NPC), which is comprised of approximately 50 unique proteins collectively known as nucleoporins. The 98 kD nucleoporin is generated through a biogenesis pathway that involves synthesis and proteolytic cleavage of a 186 kD precursor protein. This cleavage results in the 98 kD nucleoporin as well as a 96 kD nucleoporin, both of which are localized to the nucleoplasmic side of the NPC. Rat studies show that the 98 kD nucleoporin functions as one of several docking site nucleoporins of transport substrates. The human gene has been shown to fuse to several genes following chromsome translocatons in acute myelogenous leukemia (AML) and T-cell acute lymphocytic leukemia (T-ALL). This gene is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in several transcript variants; however, not all variants have been fully described. (provided by RefSeq)

Immunogen

NUP98 (AAH12906.1, 1 a.a. ~ 606 a.a) full-length human protein.

Sequence
MKLYQTPLELKLKHSTVHVDELCPLIVPNLGVAVIHDYADWVKEASGDLPEAQIVKHWSLTWTLCEALWGHLKELDSQLNEPREYIQILERRRAFSRWLSCTATPQIEEEVSLTQKNSPVEAVFSYLTGKRISEACSLAQQSGDHRLALLLSQFVGSQSVRELLTMQLVDWHQLQADSFIQDERLRIFALLAGKPVWQLSEKKQINVCSQLDWKRSLAIHLWYLLPPTASISRALSMYEEAFQNTSDSDRYACSPLPSYLEGSGCVIAEEQNSQTPLRDVCFHLLKLYSDRHYDLNQLLEPRSITADPLDYRLSWHLWEVLRALNYTHLSAQCEGVLQASYAGQLESEGLWEWAIFVLLHIDNSGIREKAVRELLTRHCQLLETPESWAKETFLTQKLRVPAKWIHEAKAVRAHMESDKHLEALCLFKAEHWNRCHKLIIRHLASDAIINENYDYLKGFLEDLAPPERSSLIQDWETSGLVYLDYIRVIEMLRHIQQVDCSGNDLEQLHIKVTSLCSRIEQIQCYSAKDRLAQSDMAKRVANLLRVVLSLHHPPDRTSDSTPDPQRVPLRLLAPHIGRLPMPEDYAMDELRSLTQSYLRELAVGSL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service