Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408807

Sigma-Aldrich

Anti-RAB32 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$583.00

$583.00


Check Cart for Availability


Select a Size

Change View
50 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 24.75 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB32(10981)

General description

Small GTP-binding proteins of the RAB family, such as RAB32, play essential roles in vesicle and granule targeting (Bao et al., 2002 [PubMed 11784320]).[supplied by OMIM

Immunogen

RAB32 (NP_006825, 1 a.a. ~ 225 a.a) full-length human protein.

Sequence
MAGGGAGDPGLGAAAAPAPETREHLFKVLVIGELGVGKTSIIKRYVHQLFSQHYRATIGVDFALKVLNWDSRTLVRLQLWDIAGQERFGNMTRVYYKEAVGAFVVFDISRSSTFEAVLKWKSDLDSKVHLPNGSPIPAVLLANKCDQNKDSSQSPSQVDQFCKEHGFAGWFETSAKDNINIEEAARFLVEKILVNHQSFPNEENDVDKIKLDQETLRAENKSQCC

Biochem/physiol Actions

RAB32 (Ras-related protein) controls intracellular lipid accumulation in hepatocytes. Absence of it promotes the expression of ATGL (adipose triglyceride lipase) and thereby causes lipolysis. RAB32 is involved in hepatic steatosis. In melanogenesis, it is involved in the transport of important enzymes, such as tyrosinase and Tyrp1 (tyrosinase related protein 1). RAB32 also interacts with and is involved in localization of leucine-rich repeat kinase 2 (LRRK2), a protein associated with Parkinson′s disease. RAB32 is hypermethylated in microsatellite-unstable colon, gastric and endometrial adenocarcinomas.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Depletion of Rab32 decreases intracellular lipid accumulation and induces lipolysis through enhancing ATGL expression in hepatocytes.
Li Q
Biochemical and Biophysical Research Communications, 471, 492-496 (2016)
LRRK2 transport is regulated by its novel interacting partner Rab32.
Waschbusch D
PLoS ONE, 9, e111632-e111632 (2014)
RAB32 hypermethylation and microsatellite instability in gastric and endometrial adenocarcinomas.
Shibata D
International Journal of Cancer. Journal International Du Cancer, 119, 801-806 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service