Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408625

Sigma-Aldrich

Anti-PTS antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ97081, PTPS

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$484.00

$484.00


Check Cart for Availability


Select a Size

Change View
100 μG
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$484.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~16.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTS(5805)

General description

The enzyme encoded by this gene catalyzes the elimination of inorganic triphosphate from dihydroneopterin triphosphate, which is the second and irreversible step in the biosynthesis of tetrahydrobiopterin from GTP. Tetrahydrobiopterin, also known as BH(4), is an essential cofactor and regulator of various enzyme activities, including enzymes involved in serotonin biosynthesis and NO synthase activity. Mutations in this gene result in hyperphenylalaninemia. (provided by RefSeq)

Immunogen

PTS (NP_000308.1, 1 a.a. ~ 145 a.a) full-length human protein.

Sequence
MSTEGGGRRCQAQVSRRISFSASHRLYSKFLSDEENLKLFGKCNNPNGHGHNYKVVVTVHGEIDPATGMVMNLADLKKYMEEAIMQPLDHKNLDMDVPYFADVVSTTENVAVYIWDNLQKVLPVGVLYKVKVYETDNNIVVYKGE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service