Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408228

Sigma-Aldrich

Anti-RHPN1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA1929, ODF5, RHOPHILIN, RHPN

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$610.00

$610.00


Estimated to ship on24 April 2025



Select a Size

Change View
50 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Estimated to ship on24 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~73.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHPN1(114822)

General description

Mouse polyclonal antibody raised against a full-length human RHPN1 protein.
The gene RHPN1 (rhophilin Rho GTPase binding protein 1) is mapped to human chromosome 8q24.3. The encoded protein belongs to the rhophilin family of Rho-binding proteins. The mRNA is strongly expressed in the testis. The protein has a Rho binding domain and a PDZ (PSD95, Dlg1 and zo-1) domain.

Immunogen

RHPN1 (NP_443156.2, 1 a.a. ~ 670 a.a) full-length human protein.

Sequence
MILEERPDGAGAGEESPRLQGCDSLTQIQCGQLQSRRAQIHQQIDKELQMRTGAENLYRATSNNRVRETVALELSYVNSNLQLLKEELEELSGGVDPGRHGSEAVTVPMIPLGLKETKELDWSTPLKELISVHFGEDGASYEAEIRELEALRQAMRTPSRNESGLELLTAYYNQLCFLDARFLTPARSLGLFFHWYDSLTGVPAQQRALAFEKGSVLFNIGALHTQIGARQDRSCTEGARRAMEAFQRAAGAFSLLRENFSHAPSPDMSAASLCALEQLMMAQAQECVFEGLSPPASMAPQDCLAQLRLAQEAAQVAAEYRLVHRTMAQPPVHDYVPVSWTALVHVKAEYFRSLAHYHVAMALCDGSPATEGELPTHEQVFLQPPTSSKPRGPVLPQELEERRQLGKAHLKRAILGQEEALRLHALCRVLREVDLLRAVISQTLQRSLAKYAELDREDDFCEAAEAPDIQPKTHQKPEARMPRLSQGKGPDIFHRLGPLSVFSAKNRWRLVGPVHLTRGEGGFGLTLRGDSPVLIAAVIPGSQAAAAGLKEGDYIVSVNGQPCRWWRHAEVVTELKAAGEAGASLQVVSLLPSSRLPSLGDRRPVLLGPRGLLRSQREHGCKTPASTWASPRPLLNWSRKAQQGKTGGCPQPCAPVKPAPPSSLKHPGWP

Application

Anti-RHPN1 antibody produced in mouse has been used in immunofluorescence, immunohistochemistry and Western blotting.[1]

Biochem/physiol Actions

In mouse model, RHPN1 (rhophilin Rho GTPase binding protein 1) is required for the maintenance of the glomerular filtration barrier. Additionally, this protein is a regulator of podocyte cytoskeleton architecture.[1]

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fujita et al.
Journal of cell science, 113 ( Pt 1), 103-112 (1999-12-11)
The small GTPase Rho; functions as a molecular switch that regulates various cellular processes such as cell adhesion, motility, gene expression and cytokinesis. We previously isolated several putative Rho; targets including rhophilin which bound selectively to the GTP-bound form of
Jeremy W Peck et al.
The Journal of biological chemistry, 277(46), 43924-43932 (2002-09-11)
The Rho GTPases regulate the actin cytoskeleton through interactions with various downstream effector molecules. Here we have identified a ubiquitously expressed human RhoA-binding protein, designated Rhophilin-2. Rhophilin-2 shows 40% amino acid similarity to human Rhophilin-1 and contains an N-terminal Rho-binding
Kyung Im Kim et al.
Basic & clinical pharmacology & toxicology, 111(5), 317-324 (2012-06-08)
Copy number variation (CNV) has been reported to be associated with chemotherapy response, which affects disease prognosis. Here, we determined the frequency of genome-wide cytogenetic CNV aberrations in Korean patients with normal karyotype (NK) acute myeloid leukaemia (AML) and tested
Mark A Lal et al.
Journal of the American Society of Nephrology : JASN, 26(3), 647-662 (2014-07-30)
Rhophilin-1 is a Rho GTPase-interacting protein, the biologic function of which is largely unknown. Here, we identify and describe the functional role of Rhophilin-1 as a novel podocyte-specific protein of the kidney glomerulus. Rhophilin-1 knockout mice were phenotypically normal at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service