Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407707

Sigma-Aldrich

Anti-ACOT13 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HT012, MGC4961, PNAS-27

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
50 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~15 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACOT13(55856)

General description

This gene encodes a member of the thioesterase superfamily. In humans, the protein co-localizes with microtubules and is essential for sustained cell proliferation. The orthologous mouse protein forms a homotetramer and is associated with mitochondria. The mouse protein functions as a medium- and long-chain acyl-CoA thioesterase. Multiple transcript variants encoding different isoforms have been found for this gene

Immunogen

ACOT13 (NP_060943.1, 1 a.a. ~ 140 a.a) full-length human protein.

Sequence
MTSMTQSLREVIKAMTKARNFERVLGKITLVSAAPGKVICEMKVEEEHTNAIGTLHGGLTATLVDNISTMALLCTERGAPGVSVDMNITYMSPAKLGEDIVITAHVLKQGKTLAFTSVDLTNKATGKLIAQGRHTKHLGN

Application

Anti-ACOT13 antibody produced in mouse is suitable for western blot anaysis.

Biochem/physiol Actions

ACOT13 (Acyl-CoA thioesterase 13) is highly involved in the regulation of hepatic lipid and glucose homeostasis. It maintains the intracellular concentration of hepatic free fatty acids (FFAs) and fatty acyl-CoAs under physiological conditions by forming a complex with phosphatidylcholine transfer protein (PC-TP). In liver, it controls the β-oxidation of fatty acids, which is highly crucial for entry of fatty acyl-CoAs into mitochondria.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Won Kang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(5), 2209-2221 (2012-02-22)
Members of the acyl-CoA thioesterase (Acot) gene family catalyze the hydrolysis of fatty acyl-CoAs, but their biological functions remain unknown. Thioesterase superfamily member 2 (Them2; synonym Acot13) is a broadly expressed mitochondria-associated Acot. Them2 was previously identified as an interacting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service