Skip to Content
Merck
All Photos(2)

Key Documents

SAB1407309

Sigma-Aldrich

Anti-FGF21 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$691.00

$691.00


Check Cart for Availability


Select a Size

Change View
50 μG
$691.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$691.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~22.3 kDa

species reactivity

human, rat

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF21(26291)

General description

FGF (fibroblast growth factor) family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined. The gene is mapped to human chromosome 19q13.33 and codes for a member of FGF superfamily. The encoded protein is produced in liver.

Immunogen

FGF21 (AAH18404.1, 1 a.a. ~ 209 a.a) full-length human protein.

Sequence
MDSDETGFEHSGLWVSVLAGLLLGACQAHPIPDSSPLLQFGGQVRQRYLYTDDAQQTEAHLEIREDGTVGGAADQSPESLLQLKALKPGVIQILGVKTSRFLCQRPDGALYGSLHFDPEACSFRELLLEDGYNVYQSEAHGLPLHLPGNKSPHRDPAPRGPARFLPLPGLPPAPPEPPGILAPQPPDVGSSDPLSMVGPSQGRSPSYAS

Biochem/physiol Actions

The members of FGF (fibroblast growth factor) family take part in cell survival and mitogenic activities. FGFs are associated with a number of physiological processes such as cell growth, morphogenesis, embryonic development, tissue repair, tumor growth and invasion. FGF21 is a hormonal factor that regulates glucose homeostasis and energy metabolism. It possesses anti-diabetic properties by mediating glucose uptake in peripheral tissues. It also exhibits autocrine effects in white adipose tissue. FGF21 is present in milk and is needed for intestinal function in the neonate. It is also associated with bone loss. FGF21 is considered hepatoprotective in nature.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A liver-bone endocrine relay by IGFBP1 promotes osteoclastogenesis and mediates FGF21-induced bone resorption.
Wang X, et al.
Cell Metabolism, 22(5), 811-824 (2015)
Growth factors and chondrogenic differentiation of mesenchymal stem cells.
Danisovic L, et al.
Tissue & cell, 44(2), 69-73 (2012)
Novel locus including FGF21 is associated with dietary macronutrient intake.
Chu A Y, et al.
Human Molecular Genetics, 22(9), 1895-1902 (2013)
Human FGF-21 is a substrate of fibroblast activation protein.
Coppage A L, et al.
PLoS ONE, 11(3), e0151269-e0151269 (2016)
Fibroblast Activation Protein Cleaves and Inactivates Fibroblast Growth Factor 21.
Dunshee DR
The Journal of Biological Chemistry, 291, 5986-5996 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service