SAB1407137
Anti-ACOT7 antibody produced in mouse
purified immunoglobulin, buffered aqueous solution
Synonym(s):
ACH1, ACT, BACH, CTE-II, LACH, LACH1, MGC1126, hBACH
Select a Size
$610.00
Select a Size
About This Item
$610.00
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~41.8 kDa
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ACOT7(11332)
General description
Immunogen
Sequence
MKLLARALRLCEFGRQASSRRLVAGQGCVGPRRGCCAPVQVVGPRADLPPCGACITGRIMRPDDANVAGNVHGGTILKMIEEAGAIISTRHCNSQNGERCVAALARVERTDFLSPMCIGEVAHVSAEITYTSKHSVEVQVNVMSENILTGAKKLTNKATLWYVPLSLKNVDKVLEVPPVVYSRQEQEEEGRKRYEAQKLERMETKWRNGDIVQPVLNPEPNTVSYSQSSLIHLVGPSDCTLHGFVHGGVTMKLMDEVAGIVAARHCKTNIVTASVDAINFHDKIRKGCVITISGRMTFTSNKSMEIEVLVDADPVVDSSQKRYRAASAFFTYVSLSQEGRSLPVPQLVPETEDEKKRFEEGKGRYLQMKAKRQGHAEPQP
Biochem/physiol Actions
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service