Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406695

Sigma-Aldrich

Anti-RFXANK antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ANKRA1, BLS, F14150_1, MGC138628, RFX-B

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
50 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~25.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RFXANK(8625)

General description

Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. This protein contains ankyrin repeats involved in protein-protein interactions. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group B. Two transcript variants encoding different isoforms have been described for this gene, with only one isoform showing activation activity. (provided by RefSeq)

Immunogen

RFXANK (NP_604389.1, 1 a.a. ~ 237 a.a) full-length human protein.

Sequence
MELTQPAEDLIQTQQTPASELGDPEDPGEEAADGSDTVVLSLFPCTPEPVNPEPDASVSSPQGSSLKHSTTLTNRQRGNEVSALPATLDCDNLVNKPDERGFTPLIWASAFGEIETVRFLLEWGADPHILAKERESALSLASTGGYTDIVGLLLERDVDINIYDWNGGTPLLYAVRGNHVKCVEALLARGADLTTEADSGYTPMDLAVALGYRKVQQVIENHILKLFQSNLVPADPE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service