Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406674

Sigma-Aldrich

Anti-SORBS2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ARGBP2, FLJ93447, KIAA0777, PRO0618

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
50 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~124.1 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORBS2(8470)

Related Categories

General description

Arg and c-Abl represent the mammalian members of the Abelson family of non-receptor protein-tyrosine kinases. They interact with the Arg/Abl binding proteins via the SH3 domains present in the carboxy end of the latter group of proteins. This gene encodes the sorbin and SH3 domain containing 2 protein. It has three C-terminal SH3 domains and an N-terminal sorbin homology (SoHo) domain that interacts with lipid raft proteins. The subcellular localization of this protein in epithelial and cardiac muscle cells suggests that it functions as an adapter protein to assemble signaling complexes in stress fibers, and that it is a potential link between Abl family kinases and the actin cytoskeleton. Alternative splicing results in multiple transcript variants encoding different isoforms. (provided by RefSeq)

Immunogen

SORBS2 (NP_066547.1, 1 a.a. ~ 1100 a.a) full-length human protein.

Sequence
MSYYQRPFSPSAYSLPASLNSSIVMQHGTSLDSTDTYPQHAQSLDGTTSSSIPLYRSSEEEKRVTVIKAPHYPGIGPVDESGIPTAIRTTVDRPKDWYKTMFKQIHMVHKPDDDTDMYNTPYTYNAGLYNPPYSAQSHPAAKTQTYRPLSKSHSDNSPNAFKDASSPVPPPHVPPPVPPLRPRDRSSTEKHDWDPPDRKVDTRKFRSEPRSIFEYEPGKSSILQHERPASLYQSSIDRSLERPMSSASMASDFRKRRKSEPAVGPPRGLGDQSASRTSPGRVDLPGSSTTLTKSFTSSSPSSPSRAKGGDDSKICPSLCSYSGLNGNPSSELDYCSTYRQHLDVPRDSPRAISFKNGWQMARQNAEIWSSTEETVSPKIKSRSCDDLLNDDCDSFPDPKVKSESMGSLLCEEDSKESCPMAWGSPYVPEVRSNGRSRIRHRSARNAPGFLKMYKKMHRINRKDLMNSEVICSVKSRILQYESEQQHKDLLRAWSQCSTEEVPRDMVPTRISEFEKLIQKSKSMPNLGDDMLSPVTLEPPQNGLCPKRRFSIEYLLEEENQSGPPARGRRGCQSNALVPIHIEVTSDEQPRAHVEFSDSDQDGVVSDHSDYIHLEGSSFCSESDFDHFSFTSSESFYGSSHHHHHHHHHHHRHLISSCKGRCPASYTRFTTMLKHERARHENTEEPRRQEMDPGLSKLAFLVSPVPFRRKKNSAPKKQTEKAKCKASVFEALDSALKDICDQIKAEKKRGSLPDNSILHRLISELLPDVPERNSSLRALRRSPLHQPLHPLPPDGAIHCPPYQNDCGRMPRSASFQDVDTANSSCHHQDRGGALQDRESPRSYSSTLTDMGRSAPRERRGTPEKEKLPAKAVYDFKAQTSKELSFKKGDTVYILRKIDQNWYEGEHHGRVGIFPISYVEKLTPPEKAQPARPPPPAQPGEIGEAIAKYNFNADTNVELSLRKGDRVILLKRVDQNWYEGKIPGTNRQGIFPVSYVEVVKKNTKGAEDYPDPPIPHSYSSDRIHSLSSNKPQRPVFTHENIQGGGEPFQALYNYTPRNEDELELRESDVIDVMEKCDDGWFVGTSRRTKFFGTFPGNYVKRL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service