Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406159

Sigma-Aldrich

Anti-PPP1R12B antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC131980, MGC87886, MYPT2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
50 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~42.57 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1R12B(4660)

General description

Myosin light chain phosphatase (MLCP) consists of three subunits- catalytic subunit, large subunit/myosin binding subunit (MBS) and small subunit (sm-M20). This gene is a multi-functional gene which encodes both MBS and sm-M20. MLCP regulates myosins and the dephosphorylation is enhanced by the presence of MBS. The sm-M20 is suggested to play a regulatory role in muscle contraction by binding to MBS. MBS is also encoded by another gene, myosin light chain phosphatase target subunit 1. sm-M20 shows higher binding affinity to this gene product than to myosin light chain phosphatase target subunit 2-MBS even though the two MBS proteins are highly similar. Although both MBSs increase the activity of MLCP, myosin light chain phosphatase target subunit 1-MBS is a more efficient activator. There are four alternatively spliced transcript variants described; two alter the MBS coding region and two alter the sm-M20 coding region of this gene. (provided by RefSeq)

Immunogen

PPP1R12B (AAH34430, 1 a.a. ~ 386 a.a) full-length human protein.

Sequence
MAELEHLGGKRAESARMRRAEQLRRWRGSLTEQEPAERRGAGRQPLTRRGSPRVRFEDGAVFLAACSSGDTDEVRKLLARGADINTVNVDGLTALHQACIDENLDMVKFLVENRANVNQQDNEGWTPLHAAASCGYLNIAEYFINHGASVGIVNSEGEVPSDLAEEPAMKDLLLEQVKKQGVDLEQSRKEEEQQMLQDARQWLNSGKIEDVRQARSGATALHVAAAKGYSEVLRLLIQAGYELNVQDYDGWTPLHAAAHWGVKEACSILAEALCDMDIRNKLGQTPFDVADEGLVEHLELLQKKQNVLRSEKETRNKLIESDLNSKIQSGFFKNKEKMLYEEETPKSQEMEEENKESSSSSSEEEEGEDEASESETEKEAVLFWPF

Biochem/physiol Actions

PPP1R12B (protein phosphatase 1 regulatory subunit 12B) is associated with childhood asthma. It is a part of the myosin phosphatase complex. The complex controls contractile responses in muscle and nonmuscle cells. PPP1R12B is upregulated in disorders, including ischaemia, atrial fibrillation, myocardial infarction and cardiomyopathy.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of PPP1R12B as a candidate gene for childhood asthma in Russians.
Maxim B Freidin et al.
Journal of genetics, 92(1), 93-96 (2013-05-04)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service