Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406111

Sigma-Aldrich

Anti-MCM5 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDC46, MGC5315, P1-CDC46

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$691.00

$691.00


Check Cart for Availability


Select a Size

Change View
50 μG
$691.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$691.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~82.3 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCM5(4174)

General description

The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation. (provided by RefSeq)

Immunogen

MCM5 (NP_006730.2, 1 a.a. ~ 734 a.a) full-length human protein.

Sequence
MSGFDDPGIFYSDSFGGDAQADEGQARKSQLQRRFKEFLRQYRVGTDRTGFTFKYRDELKRHYNLGEYWIEVEMEDLASFDEDLADYLYKQPAEHLQLLEEAAKEVADEVTRPRPSGEEVLQDIQVMLKSDASPSSIRSLKSDMMSHLVKIPGIIIAASAVRAKATRISIQCRSCRNTLTNIAMRPGLEGYALPRKCNTDQAGRPKCPLDPYFIMPDKCKCVDFQTLKLQELPDAVPHGEMPRHMQLYCDRYLCDKVVPGNRVTIMGIYSIKKFGLTTSRGRDRVGVGIRSSYIRVLGIQVDTDGSGRSFAGAVSPQEEEEFRRLAALPNVYEVISKSIAPSIFGGTDMKKAIACLLFGGSRKRLPDGLTRRGDINLLMLGDPGTAKSQLLKFVEKCSPIGVYTSGKGSSAAGLTASVMRDPSSRNFIMEGGAMVLADGGVVCIDEFDKMREDDRVAIHEAMEQQTISIAKAGITTTLNSRCSVLAAANSVFGRWDETKGEDNIDFMPTILSRFDMIFIVKDEHNEERDVMLAKHVITLHVSALTQTQAVEGEIDLAKLKKFIAYCRVKCGPRLSAEAAEKLKNRYIIMRSGARQHERDSDRRSSIPITVRQLEAIVRIAEALSKMKLQPFATEADVEEALRLFQVSTLDAALSGTLSGVEGFTSQEDQEMLSRIEKQLKRRFAIGSQVSEHSIIKDFTKQKYPEHAIHKVLQLMLRRGEIQHRMQRKVLYRLK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoqing Lv et al.
Nature cell biology, 26(9), 1585-1596 (2024-07-31)
Cyclic GMP-AMP synthase (cGAS), a cytosolic DNA sensor that initiates a STING-dependent innate immune response, binds tightly to chromatin, where its catalytic activity is inhibited; however, mechanisms underlying cGAS recruitment to chromatin and functions of chromatin-bound cGAS (ccGAS) remain unclear.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service