Skip to Content
Merck
All Photos(1)

Documents

SAB1406101

Sigma-Aldrich

Anti-MAGEA4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B, MGC21336

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~34.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGEA4(4103)

Related Categories

General description

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene. (provided by RefSeq)

Immunogen

MAGEA4 (AAH17723.1, 1 a.a. ~ 317 a.a) full-length human protein.

Sequence
MSSEQKSQHCKPEEGVEAQEEALGLVGAQAPTTEEQEAAVSSSSPLVPGTLEEVPAAESAGPPQSPQGASALPTTISFTCWRQPNEGSSSQEEEGPSTSPDAESLFREALSNKVDELAHFLLRKYRAKELVTKAEMLERVIKNYKRCFPVIFGKASESLKMIFGIDVKEVDPTSNTYTLVTCLGLSYDGLLGNNQIFPKTGLLIIVLGTIAMEGDSASEEEIWEELGVMGVYDGREHTVYGEPRKLLTQDWVQENYLEYRQVPGSNPARYEFLWGPRALAETSYVKVLEHVVRVNARVRIAYPSLREAALLEEEEGV

Biochem/physiol Actions

MAGEA4 (Melanoma antigen family A 4) induces growth in spontaneously transformed normal oral keratinocytes (NOK-SI) by blocking cell cycle at G1 phase as well as p53 mediated apoptosis. Its tumor suppressive property has also been reported. It may also act as a new marker for early detection of metastases in 160 chronic HCV (hepatitis C virus) Egyptian patients.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yousri M Hussein et al.
Medical oncology (Northwood, London, England), 29(2), 994-999 (2011-04-01)
The dissemination of hepatocellular carcinoma (HCC) cells into the circulation plays a critical role in post-operative recurrence and metastasis. Early detection of metastatic tumor cells is critical to identify HCC patients at high risk of relapse. MAGE-3 and -4 genes
R C Hillig et al.
Journal of molecular biology, 310(5), 1167-1176 (2001-08-15)
The heterotrimeric complex of the human major histocompatibity complex (MHC) molecule HLA-A*0201, beta2-microglobulin and the decameric peptide GVYDGREHTV derived from the melanoma antigen (MAGE-A4 protein has been determined by X-ray crystallography at 1.4 A resolution. MAGE-A4 belongs to a family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service