Skip to Content
Merck
All Photos(2)

Key Documents

SAB1405069

Sigma-Aldrich

Monoclonal Anti-RRM2B antibody produced in mouse

clone 6C1, ascites fluid

Synonym(s):

DKFZp686M05248, MGC102856, MGC42116, p53R2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
200 μL
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6C1, monoclonal

mol wt

antigen ~35.13 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RRM2B(50484)

Immunogen

RRM2B (NP_056528, 3 a.a. ~ 84 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DPERPEAAGLDQDERSSSDTNESEIKSNEEPLLRKSSRRFVIFPIQYPDIWKMYKQAQASFWTAEEVDLSKDLPHWNKLKAD

Physical form

Clear solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service