Skip to Content
Merck
All Photos(2)

Key Documents

SAB1404961

Sigma-Aldrich

Monoclonal Anti-EXOC7, (C-terminal) antibody produced in mouse

clone 1B7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

2-5-3p, DKFZp686J04253, EX070, EXO70, EXOC1, Exo70p, FLJ40965, FLJ46415, YJL085W

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EXOC7(23265)

General description

EXOC7 is a component of the exocyst, which is an evolutionarily conserved octameric protein complex essential for exocytosis. The exocyst targets secretory vesicles at specific domains of the plasma membrane for cell surface expansion and protein secretion (Zuo et al., 2006 [PubMed 17086175]).[supplied by OMIM

Immunogen

EXOC7 (NP_001013861, 586 a.a. ~ 684 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VFQPGVKLRDKERQIIKERFKGFNDGLEELCKIQKAWAIPDTEQRDRIRQAQKTIVKETYGAFLQKFGSVPFTKNPEKYIKYGVEQVGDMIDRLFDTSA

Application

Monoclonal Anti-EXOC7, (C-terminal) antibody produced in mouse is suitable for indirect ELISA and western blot assays.

Biochem/physiol Actions

EXOC7 (Exocyst complex component 7) is mainly involved in the exocytosis, a membrane trafficking process of intracellular protein elements such as hormones and neurotransmitters, membrane proteins and lipids to specific domains of the plasma membrane. Exocytosis plays a vital role in the cellular growth, development and cell polarity establishment. During exocytosis, it directly connects with the plasma membrane through its direct interaction with phosphatidylinositol 4,5-bisphosphate (PI(4,5)P(2)). It has also been reported that the interaction between EXOC7 and PI(4,5)P(2) is highly essential for the docking and fusion of post-Golgi secretory vesicles.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianglan Liu et al.
Molecular biology of the cell, 18(11), 4483-4492 (2007-09-01)
The exocyst is an evolutionarily conserved octameric protein complex that tethers post-Golgi secretory vesicles at the plasma membrane for exocytosis. To elucidate the mechanism of vesicle tethering, it is important to understand how the exocyst physically associates with the plasma
Shu-Chan Hsu et al.
International review of cytology, 233, 243-265 (2004-03-24)
Exocytosis is an essential membrane traffic event mediating the secretion of intracellular protein contents such as hormones and neurotransmitters as well as the incorporation of membrane proteins and lipids to specific domains of the plasma membrane. As a fundamental cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service