Skip to Content
Merck
All Photos(2)

Key Documents

SAB1404753

Sigma-Aldrich

Monoclonal Anti-KNTC1, (C-terminal) antibody produced in mouse

clone 10D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ36151, KIAA0166, ROD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10D9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KNTC1(9735)

General description

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division. Experimental evidence indicated that the encoded protein functioned in a similar manner to that of the Drosophila rough deal protein. (provided by RefSeq)

Immunogen

KNTC1 (NP_055523, 2100 a.a. ~ 2209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DPQVILKQLEEHMNTGQLAGFSHQIRSLILNNIINKKEFGILAKTKYFQMLKMHAMNTNNITELVNYLANDLSLDEASVLITEYSKHCGKPVPPDTAPCEILKMFLSGLS

Biochem/physiol Actions

KNTC1 (kinetochore associated 1) is part of the RZZ (Rod–Zw10–Zwilch) complex which is needed for proper kinetochore recruitment. Absence of KNTC1 causes checkpoint failure and early exit from mitosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A maternal effect rough deal mutation suggests that multiple pathways regulate Drosophila RZZ kinetochore recruitment.
Defachelles L, et al.
Journal of Cell Science, 128, 1204-1216 (2015)
Human Zw10 and ROD are mitotic checkpoint proteins that bind to kinetochores.
Chan GK, et al.
Nature Cell Biology, 2, 944-947 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service