Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403965

Sigma-Aldrich

Monoclonal Anti-IHH antibody produced in mouse

clone 2F3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

BDA1, HHG2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.89 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IHH(3549)

Immunogen

IHH (AAH34757, 119 a.a. ~ 217 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ESGARVALSAVRPGDRVLAMGEDGSPTFSDVLIFLDREPHRLRAFQVIETQDPPRRLALTPAHLLFTADNHTEPAARFRATFASHVQPGQYVLVAGVPG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ang Deng et al.
Molecular medicine reports, 16(6), 7987-7992 (2017-10-11)
Indian hedgehog protein (Ihh) is evolutionarily conserved and serves important roles in controlling the differentiation of progenitor cells into osteoblasts. Ihh null mutant mice exhibit a failure of osteoblast development in endochondral bone. Although studies have demonstrated that Ihh signaling
Ang Deng et al.
Experimental and therapeutic medicine, 15(1), 789-794 (2018-02-13)
The proliferative rate of chondrocytes affects bone elongation. Chondrocyte hypertrophy is required for endochondral bone formation as chondrocytes secrete factors required for osteoblast differentiation and maturation. Previous studies have demonstrated that the Indian hedgehog (Ihh) signaling pathway is a key

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service