Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403893

Sigma-Aldrich

Monoclonal Anti-GTF2E2 antibody produced in mouse

clone 2C3, ascites fluid

Synonym(s):

FE, TF2E2, TFIIE-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2C3, monoclonal

mol wt

antigen ~58.12 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTF2E2(2961)

General description

Mouse monoclonal antibody raised against a full-length recombinant GTF2E2.
The gene GTF2E2 (general transcription factor IIE subunit 2) is mapped to human chromosome 8p12.  The protein has a serine-rich region, a centrally located forkhead domain, a leucine repeat motif, a region similar to the bacterial σ factor subdomain 3, a bHLH (basic region-helix-loop-helix) motif and a bHL (basic region-helix-loop) sequence.

Immunogen

GTF2E2 (AAH30572, 1 a.a. ~ 291 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDPSLLRERELFKKRALSTPVVEKRSASSESSSSSSKKKKTKVEHGGSSGSKQNSDHSNGSFNLKALSGSSGYKFGVLAKIVNYMKTRHQRGDTHPLTLDEILDETQHLDIGLKQKQWLMTEALVNNPKIEVIDGKYAFKPKYNVRDKKALLRLLDQHDQRGLGGILLEDIEEALPNSQKAVKALGDQILFVNRPDKKKILFFNDKSCQFSVDEEFQKLWRSVTVDSMDEEKIEEYLKRQGISSMQESGPKKVAPIQRRKKPASQKKRRFKTHNEHLAGVLKDYSDITSSK

Biochem/physiol Actions

The gene GTF2E2 (general transcription factor IIE subunit 2) encodes the β subunit of transcription factor IIE (TFIIEβ). It has DNA binding ability and is essential for transcription. GTF2E2 also binds to RNA polymerase II. In patients with trichothiodystrophy, mutation in GTF2E2 is responsible for destabilization of TFIIE.

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christiane Kuschal et al.
American journal of human genetics, 98(4), 627-642 (2016-03-22)
The general transcription factor IIE (TFIIE) is essential for transcription initiation by RNA polymerase II (RNA pol II) via direct interaction with the basal transcription/DNA repair factor IIH (TFIIH). TFIIH harbors mutations in two rare genetic disorders, the cancer-prone xeroderma
Aki Tanaka et al.
Genes to cells : devoted to molecular & cellular mechanisms, 14(3), 395-405 (2009-02-13)
The human general transcription factor, TFIIE, consists of two subunits, alpha and beta. Structural analyses indicated the presence of a forkhead motif within the central region of TFIIEbeta. This motif was essential for transcription and possessed a double-stranded DNA-binding activity.
Angelica Ronald et al.
Behavior genetics, 40(1), 31-45 (2009-12-17)
Two separate genome-wide association studies were conducted to identify single nucleotide polymorphisms (SNPs) associated with social and nonsocial autistic-like traits. We predicted that we would find SNPs associated with social and non-social autistic-like traits and that different SNPs would be
Tomomichi Watanabe et al.
Molecular and cellular biology, 23(8), 2914-2926 (2003-04-01)
The general transcription factor TFIIE plays essential roles in both transcription initiation and the transition from initiation to elongation. Previously, we systematically deleted the structural motifs and characteristic sequences of the small subunit of human TFIIE (hTFIIE beta) to map

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service