Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403592

Sigma-Aldrich

Monoclonal Anti-ATP7B, (C-terminal) antibody produced in mouse

clone 3A12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

PWD, WC1, WD, WND

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$691.00

$691.00


Check Cart for Availability


Select a Size

Change View
100 μG
$691.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$691.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.45 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP7B(540)

General description

This gene is a member of the P-type cation transport ATPase family and encodes a protein with several membrane-spanning domains, an ATPase consensus sequence, a hinge domain, a phosphorylation site, and at least 2 putative copper-binding sites. This protein functions as a monomer, exporting copper out of the cells, such as the efflux of hepatic copper into the bile. Alternate transcriptional splice variants, encoding different isoforms with distinct cellular localizations, have been characterized. Mutations in this gene have been associated with Wilson disease (WD). (provided by RefSeq)

Immunogen

ATP7B (NP_000044, 1372 a.a. ~ 1465 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QLKCYKKPDLERYEAQAHGHMKPLTASQVSVHIGMDDRWRDSPRATPWDQVSYVSQVSLSSLTSDKPSRHSAAADDDGDKWSLLLNGRDEEQYI

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoru Kakuda et al.
American journal of obstetrics and gynecology, 222(1), 64-64 (2019-07-28)
Multidrug resistance is a major concern in uterine leiomyosarcoma treatment. Development of effective chemotherapies and management of drug resistance in patients is necessary. The copper efflux transporter adenosine triphosphatase copper transporting beta is a member of the P-type adenosine triphosphatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service