Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403528

Sigma-Aldrich

Monoclonal Anti-ADRA1A, (N-terminal) antibody produced in mouse

clone 5G8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADRA1C, ADRA1L1, ALPHA1AAR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~29.08 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRA1A(148)

General description

Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1A-adrenergic receptor. Alternative splicing of this gene generates four transcript variants, which encode four different isoforms with distinct C-termini but having similar ligand binding properties. (provided by RefSeq)

Immunogen

ADRA1A (NP_150646, 1 a.a. ~ 27 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MVFLSGNASDSSNCTQPPAPVNISKAI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J García-Domínguez et al.
Molecular biology of the cell, 22(8), 1227-1239 (2011-02-25)
In the vertebrate neuroepithelium, the decision to differentiate is made by neural precursors soon after mitosis, when they are apically located. This process is controlled by lateral inhibitory signals triggered by the Delta/Notch pathway. During mitosis, the capacity of neural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service