Skip to Content
Merck
All Photos(1)

Key Documents

SAB1403333

Sigma-Aldrich

Monoclonal Anti-TNRC6C antibody produced in mouse

clone 3G11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ20015, KIAA1582

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Check Cart for Availability


Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.89 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNRC6C(57690)

General description

Mouse monoclonal antibody raised against a partial recombinant TNRC6C.
TNRC6C (Trinucleotide repeat containing 6C) is a member of GW182 (Gly-Trp) family consisting of a domain similar to Drosophila GW182 (also known as Gawky), a domain rich in GW or WG repeats followed by a glutamine (Q)-rich region at the N-proximal end, a central ubiquitin-associated (UBA) domain and an RNA-binding domain termed as RRM.

Immunogen

TNRC6C (NP_061869, 181 a.a. ~ 280 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TDGPNNTNPMNSSPNPINAMQTNGLPNWGMAVGMGAIIPPHLQGLPGANGSSVSQVSGGSAEGISNSVWGLSPGNPATGNSNSGFSQGNGDTVNSALSAK

Application

Monoclonal Anti-TNRC6C antibody produced mouse is suitable for indirect ELISA.

Biochem/physiol Actions

TNRC6C (Trinucleotide repeat containing 6C) plays an important role in the microRNA repression during protein synthesis via miRNA pathway. The C-terminal RRM RNA-binding domain mediates the process of translational repression. Its GW-rich and Q-rich domain also suppress the expression for protein synthesis by binding to a reporter mRNA. In addition to translational repression, the members of the GW182 family, also participate in the deadenylation and decay of miRNA by interacting with argonaute proteins.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Latifa Zekri et al.
The EMBO journal, 32(7), 1052-1065 (2013-03-07)
GW182 family proteins interact with Argonaute proteins and are required for the translational repression, deadenylation and decay of miRNA targets. To elicit these effects, GW182 proteins interact with poly(A)-binding protein (PABP) and the CCR4-NOT deadenylase complex. Although the mechanism of
Jakob T Zipprich et al.
RNA (New York, N.Y.), 15(5), 781-793 (2009-03-24)
Proteins of the GW182 family play an important role in the execution of microRNA repression in metazoa. They interact directly with Argonaute proteins, components of microRNPs, and also form part of P-bodies, structures implicated in translational repression and mRNA degradation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service