Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403129

Sigma-Aldrich

Monoclonal Anti-SULF1, (C-terminal) antibody produced in mouse

clone 1A4, ascites fluid

Synonym(s):

FLJ30905, FLJ38022, FLJ41750, HSULF-1, KIAA1077, SULF-1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$691.00

$691.00


Check Cart for Availability


Select a Size

Change View
200 μL
$691.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$691.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1A4, monoclonal

mol wt

antigen ~36.23 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULF1(23213)

General description

Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 [PubMed 16192265]).[supplied by OMIM

Immunogen

SULF1 (NP_055985.1, 780 a.a. ~ 871 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RTVNETHNFLFCEFATGFLEYFDMNTDPYQLTNTVHTVERGILNQLHVQLMELRSCQGYKQCNPRPKNLDVGNKDGGSYDLHRGQLWDGWEG

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service