Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402990

Sigma-Aldrich

Monoclonal Anti-FCN3 antibody produced in mouse

clone 4B4, ascites fluid

Synonym(s):

FCNH, HAKA1, MGC22543

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4B4, monoclonal

mol wt

antigen ~57.79 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCN3(8547)

General description

Ficolins are a group of proteins which consist of a collagen-like domain and a fibrinogen-like domain. In human serum, there are two types of ficolins, both of which have lectin activity. The protein encoded by this gene is a thermolabile beta-2-macroglycoprotein found in all human serum and is a member of the ficolin/opsonin p35 lectin family. The protein, which was initially identified based on its reactivity with sera from patients with systemic lupus erythematosus, has been shown to have a calcium-independent lectin activity. The protein can activate the complement pathway in association with MASPs and sMAP, thereby aiding in host defense through the activation of the lectin pathway. Alternative splicing occurs at this locus and two variants, each encoding a distinct isoform, have been identified. (provided by RefSeq)

Immunogen

FCN3 (AAH20731, 1 a.a. ~ 288 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDLLWILPSLWLLLLGGPACLKTQEHPSCPGPRELEASKVVLLPSCPGAPGSPGEKGAPGPQGPPGPPGKMGPKGEPGPRNCRELLSQGATLSGWYHLCLPEGRALPVFCDMDTEGGGWLVFQRRQDGSVDFFRSWSSYRAGFGNQESEFWLGNENLHQLTLQGNWELRVELEDFNGNRTFAHYATFRLLGEVDHYQLALGKFSEGTAGDSLSLHSGRPLTTYDADHDSSNSNCAVIVHGAWWYASCYRSNLNGRYAVSEAAAHKYGIDWASGRGVGHPYRRVRMMLR

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service