Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402583

Sigma-Aldrich

Monoclonal Anti-VGLL1, (N-terminal) antibody produced in mouse

clone 3C7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

TDU, VGL1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Check Cart for Availability


Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VGLL1(51442)

General description

Vestigial like family member 1 (VGLL1) is a transcriptional coactivator which is related to the Drosophila transcriptional co-activator Vg. The gene encoding it is localized on human chromosome Xq26.3.

Immunogen

VGLL1 (NP_057351.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEEMKKTAIRLPKGKQKPIKTEWNSRCVLFTYFQGDISSVVDEHFSRALSNIKSPQELTPSSQSEGVMLKNDDSMSPNQWRYSSPWTKPQPEVPVTNRAA

Biochem/physiol Actions

Vestigial like family member 1 (VGLL1) may function as a coactivator for TEA domain family of transcription factors (TEFs). It has been associated with breast cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TONDU (TDU), a novel human protein related to the product of vestigial (vg) gene of Drosophila melanogaster interacts with vertebrate TEF factors and substitutes for Vg function in wing formation.
Vaudin P
Development (1999)
VGLL1 expression is associated with a triple-negative basal-like phenotype in breast cancer.
Castilla MA
Endocrine-Related Cancer (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service