Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402559

Sigma-Aldrich

Monoclonal Anti-PVRL3 antibody produced in mouse

clone 1D1, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$718.00

$718.00


Check Cart for Availability


Select a Size

Change View
100 μG
$718.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$718.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.99 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PVRL3(25945)

General description

Poliovirus receptor-related 3 (PVRL3), popularly known as nectin cell adhesion molecule 3 (NECTIN3), is expressed in various cells. It is part of the nectin family of proteins, which are cell adhesion molecules that modulate adherens junction formation. PVRL3 is a membrane protein with three extracellular immunoglobulin domains. The gene encoding it is localized on human chromosome 3q13.13.

Immunogen

PVRL3 (NP_056295, 59 a.a. ~ 166 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PIIVEPHVTAVWGKNVSLKCLIEVNETITQISWEKIHGKSSQTVAVHHPQYGFSVQGEYQGRVLFKNYSLNDATITLHNIGFSDSGKYICKAVTFPLGNAQSSTTVTV

Application

Monoclonal Anti-PVRL3 antibody produced in mouse has been used for immunohistochemistry.

Biochem/physiol Actions

Poliovirus receptor-related 3 (PVRL3) or nectin cell adhesion molecule 3 (NECTIN3) forms heterotypic adhesions with PVR, PVRL1 and PVRL2. The protein is involved in lymphocyte and monocyte extravasation. It also associates with afadin. PVRL3 has a role in the development of mammalian lens and ciliary body.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nectin4/PRR4, a new afadin-associated member of the nectin family that trans-interacts with nectin1/PRR1 through V domain interaction.
Reymond N
The Journal of Biological Chemistry (2001)
Identification of an epithelial cell receptor responsible for Clostridium difficile TcdB-induced cytotoxicity
Michelle E. LaFrance
Proceedings of the National Academy of Sciences of the USA (2015)
The expression of the Nectin complex in human breast cancer and the role of Nectin-3 in the control of tight junctions during metastasis.
Martin TA
PLoS ONE (2013)
Nectin-3 (CD113) interacts with Nectin-2 (CD112) to promote lymphocyte transendothelial migration.
Devilard E
PLoS ONE (2013)
The cell adhesion gene PVRL3 is associated with congenital ocular defects.
Lachke SA
Human Genetics (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service