Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402549

Sigma-Aldrich

Monoclonal Anti-ZHX3 antibody produced in mouse

clone 1D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA0395, TIX1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Estimated to ship on03 May 2025



Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Estimated to ship on03 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.34 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ZHX3(23051)

General description

This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor. (provided by RefSeq)

Immunogen

ZHX3 (NP_055850.1, 863 a.a. ~ 955 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLET

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service