Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402485

Sigma-Aldrich

Monoclonal Anti-RAPGEF5 antibody produced in mouse

clone 2E4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

GFR, KIAA0277, MR-GEF, REPAC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Estimated to ship on25 May 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~74.95 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAPGEF5(9771)

General description

Members of the RAS (see HRAS; MIM 190020) subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RAPGEF5, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation (Rebhun et al., 2000 [PubMed 10934204]).[supplied by OMIM

Immunogen

RAPGEF5 (AAH39203, 1 a.a. ~ 444 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGSSRLRVFDPHLERKDSAAALSDRELPLPTFDVPYFKYIDEEDEDDEWSSRSQSSTEDDSVDSLLSDRYVVVSGTPEKILEHLLNDLHLEEVQDKETETLLDDFLLTYTVFMTTDDLCQALLRHYSAKKYQGKEENSDVPRRKRKVLHLVSQWIALYKDWLPEDEHSKMFLKTIYRNVLDDVYEYPILEKELKEFQKILGMHRRHTVDEYSPQKKNKALFHQFSLKENWLQHRGTVTETEEIFCHVYITEHSYVSVKAKVSSIAQEILLCSQLGKRVQLVKKFIKIAAHCKAQRNLNSFFAIVMGLNTASVSRLSQTWEKIPGKFRKLFSELESLTDPSLNHKAYRDAFKKMKPPKIPFVPLLLKDVTFIHEGNKTFLDNLVNFEKLHMIADTVRTLRHCRTNQFGDLSPKEHQELKSYVNHLYVIDSQQALFELSHRIEPRV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service