Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402161

Sigma-Aldrich

Monoclonal Anti-CSNK1E antibody produced in mouse

clone 2E1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HCKIE, MGC10398

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Check Cart for Availability


Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
proximity ligation assay: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSNK1E(1454)

General description

The protein encoded by this gene is a serine/threonine protein kinase and a member of the casein kinase I protein family, whose members have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. The encoded protein is found in the cytoplasm as a monomer and can phosphorylate a variety of proteins, including itself. This protein has been shown to phosphorylate period, a circadian rhythm protein. Two transcript variants encoding the same protein have been found for this gene. (provided by RefSeq)

Immunogen

CSNK1E (NP_689407, 2 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ELRVGNKYRLGRKIGSGSFGDIYLGANIASGEEVAIKLECVKTKHPQLHIESKFYKMMQGGVGIPSIKWCGAEGDYNVMVMELLGPSLEDLFNFCSRKF

Biochem/physiol Actions

Casein kinase 1 ε (CSNK1E) phosphorylates proteins of the Wnt signaling cascade. It modulates tumor growth and cell division in salivary gland cancer, pancreatic and colon adenocarcinoma cells. CSNK1E phosphorylates dopamine- and cAMP-regulated neuronal phosphoprotein (DARPP-32) and modulates dopamine receptor signaling. It also interacts with proteins involved in circadian rhythm.[1]

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of genes associated with tumorigenesis of meibomian cell carcinoma by microarray analysis.
Kumar A, et. al
Genomics, 90(5), 559-566 (2007)
Jose Luis Lopez-Guerra et al.
Oncotarget, 6(30), 30343-30356 (2015-09-04)
Reliable biological markers that predict breast cancer (BC) outcomes after multidisciplinary therapy have not been fully elucidated. We investigated the association between casein kinase 1 epsilon (CK1ε) and the risk of recurrence in patients with BC. Using 168 available tumor
Association between a casein kinase 1 e gene polymorphism and schizophrenia in a Chinese Han population.
Huang Y
Journal of Molecular Neuroscience, 47(3), 470-474 (2012)
Dopaminergic pathway polymorphisms and heroin addiction: further support for association of CSNK1E variants.
Levran O
Pharmacogenomics, 15(16), 2001-2009 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service