Skip to Content
Merck
All Photos(4)

Key Documents

SAB1401244

Sigma-Aldrich

Monoclonal Anti-MGAT5 antibody produced in mouse

clone 3E9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

GNT-V, GNT-VA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$548.00

$548.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 μG
$548.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$548.00


Estimated to ship on31 May 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MGAT5(4249)

General description

This gene encodes mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, a glycosyltransferase involved in the synthesis of protein-bound and lipid-bound oligosaccharides. Alterations of the oligosaccharides on cell surface glycoproteins cause significant changes in the adhesive or migratory behavior of a cell. Increase in the encoded protein′s activity may correlate with the progression of invasive malignancies. (provided by RefSeq)

Immunogen

MGAT5 (NP_002401, 642 a.a. ~ 739 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LAEPGQSCKQVCQESQLICEPSFFQHLNKDKDMLKYKVTCQSSELAKDILVPSFDPKNKHCVFQGDLLLFSCAGAHPRHQRVCPCRDFIKGQVALCKD

Biochem/physiol Actions

MGAT5 (mannosyl (α-1,6-)-glycoprotein β-1,6-N-acetylglucosaminyltransferase) is involved in the biosynthesis of N-linked glycoproteins. It catalyzes the formation of β
1,6-branched N-glycans by N-acetyl-d-glucosamine transfer. MGAT5 plays a significant role in invasion and metastasis of various cancer, including glioma and hepatocellular carcinoma. It is also known to be associated with multiple sclerosis and liver fibrosis. Increased radiosensitivity of cancer cells is observed upon MGAT5 inhibition.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oligosaccharide modification by N-acetylglucosaminyltransferase-V in macrophages are involved in pathogenesis of bleomycin-induced scleroderma.
Kato A
Experimental Dermatology, 24(8), 585-590 (2015)
Effect of GnT-V knockdown on the proliferation, migration and invasion of the SMMC7721/R human hepatocellular carcinoma drug-resistant cell line.
Li B
Molecular Medicine Reports, 13(1), 469-476 (2016)
N-acetylglucosaminyltransferase V modulates radiosensitivity and migration of small cell lung cancer through epithelial-mesenchymal transition.
Huang C
FEBS Journal, 282(22), 4295-4306 (2015)
N-acetylglucosaminyltransferase V inhibits the invasion of trophoblast cells by attenuating MMP2/9 activity in early human pregnancy.
Deng Q
Placenta, 36(11), 1291-1299 (2015)
Hexosamine-Induced TGF-? Signaling and Osteogenic Differentiation of Dental Pulp Stem Cells Are Dependent on N-Acetylglucosaminyltransferase V.
Chen YJ
BioMed Research International, 2015:924397, 1-11 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service