Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401241

Sigma-Aldrich

Anti-MEF2C antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$548.00

$548.00


Check Cart for Availability


Select a Size

Change View
50 μG
$548.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$548.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEF2C(4208)

Immunogen

MEF2C (AAH26341.1, 1 a.a. ~ 469 a.a) full-length human protein.

Sequence
MGRKKIQITRIMDERNRQVTFTKRKFGLMKKAYELSVLCDCEIALIIFNSTNKLFQYASTDMDKVLLKYTEYNEQHESRTNSDIVETLRKKGLNGCDSPDPDADDSVGHSPESEDKYRKINEDIDLMISRQRLCAVPPPNFEMPVSIPVSSHNSLVYSNPVSSLGNPNLLPLAHPSLQRNSMSPGVTHRPPSAGNTGGLMGGDLTSGAGTSAGNGYGNPRNSPGLLVSPGNLNKNMQAKSPPPMNLGMNNRKPDLRVLIPPGSKNTMPSVSEDVDLLLNQRINNSQSAQSLATPVVSVATPTLPGQGMGGYPSAISTTYGTEYSLSSADLSSLSGFNTASALHLGSVTGWQQQHLHNMQPSALSQLGACTSTHLSQSSNLSLPSTQSLNIKSEPVSPPRDRTTTPSRYPQHTRHEAGRSPVDSLSSCSSSYDGSDREDHRNEFHSPIGLTRPSPDERESPSVKRMRLSE

Biochem/physiol Actions

MEF2C (myocyte enhancer factor 2C) is a cardiac and smooth muscle-specific transcription factor. It positively regulates the transcriptional activation induced by myocardin. Thus, it coordinates the smooth muscle cell differentiation. MEF2C is associated with nuclear factor of activated T-cells (NFAT) and upregulation of the gene is observed in mitral regurgitation. In retina, the MEF2C regulates the expression of photoreceptor gene.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retinal expression and localization of Mef2c support its important role in photoreceptor gene expression.
Wolf A
Biochemical and Biophysical Research Communications, 483(1), 346-351 (2017)
Transcriptome and Metabolome Analyses in Exogenous FABP4- and FABP5-Treated Adipose-Derived Stem Cells.
Yamamoto T
PLoS ONE, 11(12), 1-19 (2016)
ABCG1 and ABCG4 Suppress ?-Secretase Activity and Amyloid ? Production.
Sano O
PLoS ONE, 11(5), 1-19 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service