Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400224

Sigma-Aldrich

Anti-PRPSAP2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MGC117304, Anti-MGC126719, Anti-MGC126721, Anti-PAP41

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$484.00

$484.00


Check Cart for Availability


Select a Size

Change View
50 μG
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$484.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRPSAP2(5636)

General description

The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS. (provided by RefSeq)

Immunogen

PRPSAP2 (NP_002758.1, 1 a.a. ~ 369 a.a) full-length human protein.

Sequence
MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDD

Biochem/physiol Actions

PRPSAP2 (Phosphoribosyl pyrophosphate synthetase-associated protein 2) is highly involved in the biogenesis of purine and pyrimidine nucleotids, histidine and tryptophan, and NAD (Nicotinamide adenine dinucleotide). It acts as a primary substrate during nucleotide synthesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O H Minchenko et al.
Ukrainian biochemical journal, 86(6), 74-83 (2015-03-31)
Activation of pentose phosphate pathway is an important factor of enhanced cell proliferation and tumor growth. Phosphoribosyl pyrophosphate synthetase (PRPS) is a key enzyme of this pathway and plays a central role in the synthesis of purines and pyrimidines. Hypoxia
Assignment of the human phosphoribosylpyrophosphate synthetase-associated protein 41 gene (PRPSAP2) to 17p11.2-p12.
R Katashima et al.
Genomics, 54(1), 180-181 (1998-11-10)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service