Skip to Content
Merck
All Photos(1)

Key Documents

SAB1307046

Sigma-Aldrich

ANTI-APCDD1(N-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

APCDD1, Adenomatosis polyposis coli down-regulated 1 protein, DRAPC1, Protein APCDD1

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
$698.00

$698.00


Check Cart for Availability


Select a Size

Change View
400 μL
$698.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$698.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58797 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APCDD1(147495)

General description

Adenomatosis polyposis coli down-regulated 1 (APCDD1) encodes a membrane bound glycoprotein. Apcdd1 is predominantly expressed in epidermal and dermal compartments of human hair follicles. In human chromosome, the gene APCDD1 is localized on 18p11.22.

Biochem/physiol Actions

Adenomatosis polyposis coli down-regulated 1 (APCDD1) inhibits wnt (wingless/integrated) signalling and promotes human hair growth. Mutations in APCDD1 results in a rare autosomal dominant hereditary hypotrichosis, which is characterized by hair loss due to hair follicle miniaturization. APCDD1 is directly regulated by β-catenin/T-cell factor 4 (TCF4). The high expression of Apcdd1 in colon cancer cells promotes its proliferation. APCDD1 is essential for adipocyte differentiation. Downregulation of APCDD1 in diet induced obesity impairs adipocyte differentiation and causes obesity related metabolic diseases.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel role for Wnt inhibitor APCDD1 in adipocyte differentiation: implications for diet-induced obesity
Yiew NKH, et al.
The Journal of Biological Chemistry, jbc-M116 (2017)
Yutaka Shimomura et al.
Nature, 464(7291), 1043-1047 (2010-04-16)
Hereditary hypotrichosis simplex is a rare autosomal dominant form of hair loss characterized by hair follicle miniaturization. Using genetic linkage analysis, we mapped a new locus for the disease to chromosome 18p11.22, and identified a mutation (Leu9Arg) in the adenomatosis
Meiko Takahashi et al.
Cancer research, 62(20), 5651-5656 (2002-10-18)
To clarify the molecular mechanisms of human carcinogenesis associated with abnormal beta-catenin/T-cell factor (Tcf) signaling, we have been using cDNA microarrays to search for genes whose expression is significantly altered after introduction of wild-type APC into SW480 colon cancer cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service