SAB1306800
ANTI-PCDH18(C-TERMINAL) antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
KIAA1562, PCDH18, Protocadherin-18
Select a Size
$616.00
Select a Size
About This Item
$616.00
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
126149 Da
species reactivity
mouse
technique(s)
western blot: 1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PCDH18(54510)
1 of 4
This Item | HPA017976 | SAB1304925 | HPA053921 |
---|---|---|---|
biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form IgG fraction of antiserum | antibody form affinity isolated antibody |
mol wt 126149 Da | mol wt - | mol wt 136064 Da | mol wt - |
Gene Information human ... PCDH18(54510) | Gene Information human ... PCDH18(54510) | Gene Information human ... PCDH9(5101) | Gene Information human ... PCDHB1(29930) |
form buffered aqueous solution | form buffered aqueous glycerol solution | form buffered aqueous solution | form buffered aqueous glycerol solution |
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service