Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306078

Millipore

ANTI-FLAG TAG (DYKDDDDK) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

all

concentration

(see Certificate of Analysis)

technique(s)

western blot: 1:1000

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

General description

FLAG is an octapeptide containing the following amino acid sequence Asp-Try-Lys-Asp-Asp-Asp-Asp-Lys.

Application

ANTI-FLAG TAG (DYKDDDDK) antibody produced in rabbit has been used in western blotting, (1:5000), (1:10000)

Biochem/physiol Actions

FLAG is a widely used epitope tag for immunoaffinity purification of fusion proteins.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunwang Xu et al.
Nature communications, 10(1), 5492-5492 (2019-12-04)
Gemcitabine is the first-line treatment for locally advanced and metastatic gallbladder cancer (GBC), but poor gemcitabine response is universal. Here, we utilize a genome-wide CRISPR screen to identify that loss of ELP5 reduces the gemcitabine-induced apoptosis in GBC cells in
Zhe Wang et al.
JCI insight, 4(1) (2019-01-11)
Deposition of amyloid-β protein (Aβ) to form neuritic plaques is the characteristic neuropathology of Alzheimer's disease (AD). Aβ is generated from amyloid precursor protein (APP) by β- and γ-secretase cleavages. BACE1 is the β-secretase and its inhibition induces severe side
Renxu Chang et al.
Nature communications, 9(1), 3486-3486 (2018-08-30)
Loss of WW domain-containing oxidoreductase (Wwox) expression has been observed in breast cancer (BC). However, its regulatory effects are largely unknown, especially in triple-negative breast cancer (TNBC). Herein, gene expression profiling revealed that JAK/STAT3 pathway was one of the most
Feiran Zhang et al.
Human molecular genetics, 27(22), 3936-3950 (2018-08-15)
N6-methyladenosine (m6A) is the most prevalent internal modification of mammalian messenger RNAs (mRNAs) and long non-coding RNAs. The biological functions of this reversible RNA modification can be interpreted by cytoplasmic and nuclear 'm6A reader' proteins to fine-tune gene expression, such
Yajie Gao et al.
STAR protocols, 3(3), 101596-101596 (2022-08-06)
Here, we provide an optimized RNA-induced silencing complex (RISC) assembly and cleavage protocol in vitro without using radiolabeled RNA. The protocol is useful to characterize the biochemical properties of the RISC. We describe the preparation of RNA probes, the target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service