Ubiquitin specific peptidase 51 (USP51) encodes a deubiquitylating enzyme. USP51 is predominantly expressed in prostate, brain, lung, aorta and kidney. In human chromosome, the gene USP51 is localized on Xp11.
Biochem/physiol Actions
Ubiquitin specific peptidase 51 (USP51) in response to DNA damage deubiquitylates histone H2A at Lys13,15 (H2AK13,15ub) present at DNA double-strand breaks (DSBs) to maintain genome integrity. High expression of USP51 suppresses ionizing radiations induced damage on p53-binding protein 1(53BP1) and breast cancer type 1 susceptibility protein (BRCA1). Cells depleted with USP51 show an increase in G1 phase cells and decrease in S-phase cells and hence it is required for cell cycle progression. USP51 deubiquitinase zinc finger E-Box binding homeobox 1)(ZEB1) and stabilizes its expression, especially in breast tumour cells.
Physical form
Supplied in PBS with 0.09% (W/V) sodium azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal transduction and targeted therapy, 5(1), 25-25 (2020-04-17)
Tumor metastasis is the most common cause of cancer-related deaths, yet it remains poorly understood. The transcription factor zinc-finger E-box binding homeobox 1 (ZEB1) is involved in the epithelial-to-mesenchymal transition (EMT) and plays a pivotal role in tumor metastasis. However
Cancer communications (London, England), 43(7), 765-787 (2023-06-30)
Programmed death ligand 1 (PD-L1) has been demonstrated to facilitate tumor progression and therapeutic resistance in an immune-independent manner. Nevertheless, the function and underlying signaling network(s) of cancer cell-intrinsic PD-L1 action remain largely unknown. Herein, we sought to better understand
International journal of biological sciences, 18(5), 2186-2201 (2022-03-29)
TNBC is characterized by high incidence of visceral metastasis and lacks effective clinical targets. This study aims to delineate the molecular mechanisms of SENP1 in TNBC invasion and metastasis. By using IHC to test the SENP1 expression in TNBC tissues
USP51 deubiquitylates H2AK13, 15ub and regulates DNA damage response
Wang Z, et al.
Genes & Development, 30(8), 946-959 (2016)
USP51 promotes deubiquitination and stabilization of ZEB1
Zhou Z, et al.
American Journal of Cancer Research, 7(10), 2020-2020 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.